Learn More
Azlon™ Class B PMP Volumetric Flask
Product Code. 12453621
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
Unit Size:
Each
Product Code. 12453621
Brand: Azlon™ FDF138N
Only null left
Add to basket
Add to basket
Manufactured from transparent PMP provides clear easy to read calibration line
    • Calibrated to ISO 1042, class B tolerances
    • Supplied with polypropylene stopper
TRUSTED_SUSTAINABILITY
Specifications
Color Transparent
Capacity (Metric) 1000 mL
Material PMP
Height (Metric) 310 mm (with stopper)
No. per Pack 1
Tolerance ±0.8 mL
Autoclavable Autoclavable
Class B
Closure Type Stopper