Learn More
Azlon™ Oval Stir Magnets
Product Code. 15798067
Change view
Click to view available options
Length (Metric):
10 mm
15 mm
Diameter (Metric):
5 mm
6 mm
Unit Size:
Pack of 2
Product Code. Length (Metric) Diameter (Metric) unitSize
15798067 15 mm 6 mm Pack of 2
15788067 10 mm 5 mm Pack of 2
2 options
Product Code. 15798067
Brand: Azlon SWN519
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Azlon™ Oval Stir Magnets are resistant to virtually all chemicals and are supplied in polarity aligned packs to ensure maximum coupling strength.
  • For efficient stirring
  • Oval shaped
TRUSTED_SUSTAINABILITY
Specifications
Coating PTFE
Diameter (Metric) 6 mm
Length (Metric) 15 mm
Quantity 2
Shape Oval
Type Magnetic Stir Bar