Learn More
Azlon™ Oval Stir Magnets
Description
- For efficient stirring
- Oval shaped
Specifications
Specifications
| Diameter (Metric) | 5 mm |
| Length (Metric) | 10 mm |
| Quantity | 2 |
| Shape | Oval |