Learn More
Azlon™ Industrial HDPE Funnel
Product Code. 12044837
Change view
Click to view available options
Diameter (Metric)Top:
80mm
100mm
120mm
140mm
180mm
230mm
245mm
290mm
380mm
Diameter (Metric) Stem:
9mm
11mm
12mm
13mm
17mm
21mm
24mm
27mm
36mm
Height (Metric):
80 mm
102 mm
121 mm
141 mm
181 mm
226 mm
239 mm
277 mm
351 mm
Unit Size:
Each
Pack of 10
Pack of 5
Product Code. Diameter (Metric)Top Diameter (Metric) Stem Height (Metric) unitSize
12044837 140mm 13mm 141 mm Pack of 5
11363664 100mm 11mm 102 mm Pack of 10
11373664 120mm 12mm 121 mm Pack of 10
11383664 180mm 17mm 181 mm Pack of 5
10575115 230mm 21mm 226 mm Pack of 5
11393664 245mm 24mm 239 mm Each
10504355 290mm 27mm 277 mm Each
12090773 380mm 36mm 351 mm Each
12511456 80mm 9mm 80 mm Pack of 10
9 options
Product Code. 12044837
Brand: Azlon FWC108
Only null left
Add to basket
Add to basket
Constructed of Polyethylene and unbreakable in normal use. Azlon™ Industrial HDPE Funnels are suitable for food contact applications in accordance with EU and USA regulations. Feature straight sided upper rims to prevent spillage.
  • Rigid industrial funnels in polyethylene
  • Unbreakable in normal use
  • Feature straight sided upper rims to prevent spillage
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric)Top 140mm
Capacity (Metric) 750 mL
Diameter (Metric) Stem 13mm
Material HDPE
Height (Metric) 141 mm