Learn More
Azlon™ Flow Indicator
Click to view available options
Case Material:
PMP
Unit Size:
Each
Description
- Transparent robust moulding with a colored paddle wheel
- Operates at low flow rates. (The impeller starts to turn at 150mL/min)
- Stepped side arms are tapered to accept tubing from 6mm to 11mm I. D.
Specifications
Specifications
| Type | Flow Indicator |
| Case Material | PMP |
| Color | Transparent |
| Flow Rate Range | 2 mL/sec. |
| Internal Part Material | Polystyrene |
| Length (Metric) | 90 mm |
| Temperature (Metric) | -180 to +145°C |