Learn More
Azlon™ Ampoule Breaker
Product Code. p-8001778
Click to view available options
For Use With:
1-4 mL Ampoule
5-10 mL Ampoule
Unit Size:
Pack of 1000
Product Code. 12732266
Brand: Azlon AWY700
Only null left
Add to basket
Add to basket
Break glass ampoules safely with easy and safe disposable of the broken glass . The Azlon™ ampoule breaker comes in a choice of sizes.
  • Enables safe and easy breaking of glass ampoules
  • Broken ampoule top is retained for safe disposal
  • Choice of size to suit all common glass ampoules
TRUSTED_SUSTAINABILITY
Specifications
Color Natural
Product Type Ampule Breaker
Material HDPE
For Use With 1-4 mL Ampoule