Learn More
Azlon™ Ampoule Breaker
Product Code. 12732266
Change view
Click to view available options
For Use With (Equipment):
1-4 mL Ampoule
5-10 mL Ampoule
Unit Size:
Pack of 1000
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
12732266 1-4 mL Ampoule Pack of 1000
12351168 5-10 mL Ampoule Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12732266 Supplier Azlon Supplier No. AWY700
  • ONLINE EXCLUSIVE PRICE
    5465.00 NOK
    4603.75 NOK / Pack of 1000
    Save 861.25 NOK
    15% Off
Estimated Shipment: 21-12-2026
Break glass ampoules safely with easy and safe disposable of the broken glass . The Azlon™ ampoule breaker comes in a choice of sizes.
  • Enables safe and easy breaking of glass ampoules
  • Broken ampoule top is retained for safe disposal
  • Choice of size to suit all common glass ampoules
Specifications
Color Natural
Product Type Ampule Breaker
Material HDPE
For Use With (Equipment) 1-4 mL Ampoule