Learn More
Azlon™ Ampoule Breaker
Description
- Enables safe and easy breaking of glass ampoules
- Broken ampoule top is retained for safe disposal
- Choice of size to suit all common glass ampoules
Specifications
Specifications
| Color | Natural |
| Product Type | Ampule Breaker |
| Material | HDPE |
| For Use With | 5-10 mL Ampoule |