missing translation for 'onlineSavingsMsg'
Learn More

Azithromycin, MedChemExpress

Product Code. 16473397
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16473397 50 mg 50mg
16427346 100 mg 100mg
16481266 200 mg 200mg
16491866 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16473397 Supplier Medchem Express Supplier No. HY17506

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Azithromycin is a macrolide antibiotic useful for the treatment of a number of bacterial infections.

Azithromycin is a macrolide antibiotic useful for the treatment of a number of bacterial infections.

Specifications

Chemical Name or Material Azithromycin
Physical Form Solid
CAS 83905-01-5
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C38H72N2O12
For Use With (Application) COVID-19-anti-virus
Synonym CP 62993
Solubility Information DMSO : ≥ 100 mg/mL (133.51 mM)
SMILES C[C@@H]([C@@H]([C@@](C(O[C@@H]([C@@](C)(O)[C@@H]1O)CC)=O)([H])C)O[C@@](O[C@@H](C)[C@@H]2O)([H])C[C@@]2(C)OC)[C@H]([C@](O)(C[C@H](CN([C@@H]1C)C)C)C)O[C@@](O[C@H](C)C[C@@H]3N(C)C)([H])[C@@H]3O
Molecular Weight (g/mol) 748.98
Formula Weight 748.98
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.