Learn More
Azilsartan-d5, MedChemExpress
Product Code. 16428847
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Product Code. 16428847
Brand: Medchem Express HY14914S
MedChemExpress Azilsartan D5 (TAK-536 D5) is the deuterium labeled Azilsartan(TAK-536), which is a specific and potent angiotensin II type 1 receptor antagonist.
Azilsartan D5 (TAK-536 D5) is the deuterium labeled Azilsartan(TAK-536), which is a specific and potent angiotensin II type 1 receptor antagonist.
Specifications
Chemical Name or Material Azilsartan-d5
CAS 1346599-45-8
Purity Grade Notes Research
Physical Form Solid
Molecular Formula C25H15D5N4O5
Color White
Synonym TAK-536-d5
SMILES OC(C1=CC=CC2=C1N(C(OC(C([2H])([2H])[2H])([2H])[2H])=N2)CC(C=C3)=CC=C3C4=CC=CC=C4C5=NC(ON5)=O)=O
Molecular Weight (g/mol) 461.48
Quantity 5 mg
Formula Weight 461.48
Percent Purity 98.0%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.