missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Scharlab Azide Dextrose Broth
Description
- COMPF
- Medium for the detection and enumeration of enterococci in water.
- Azide Dextrose Broth according to Rothe
Specifications
Specifications
| Form | Solid |
| Media Type | Azide Dextrose Broth |
| Product Type | Agar |
| Target Organism Class | Gram-positive Cocci |
| Formulation | Meat Peptone 10.0 g/L, Casein Peptone 10.0 g/L, Dextrose 5.0 g/L, Sodium Chloride 5.0 g/L, Dipotassium Hydrogen Phosphate 2.7 g/L, Potassium Dihydrogen Phosphate 2.7 g/L, Sodium Azide 0.2 g/L |
| For Use With (Application) | Detection and Enumeration of Enterococci aq. Soln. |
| Incubation Conditions | 36°C ± 2.0°C |
| Incubation Time | 44 ±4 hr. |
| pH | 7.0 ±0.2 |
| Quantity | 500 g |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?