Learn More
AZI2/NAP1 Rabbit anti-Human, Polyclonal, Proteintech
Product Code. 16874014 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16874014 150 μL 150µL
16884014 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16874014 Supplier Proteintech Supplier No. 150421AP150UL
Rabbit Polyclonal Antibody
AZI2, or NAP1, contributes to the activation of NFKB (Fujita et al., 2003 ).
Specifications
Antigen AZI2/NAP1
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.41 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene AZI2
Gene Accession No. Q9H6S1
Gene Alias 5 azacytidine induced 2, AZ2, AZI2, Nak associated protein 1, NAP1, TILP
Gene Symbols AZI2
Host Species Rabbit
Immunogen AZI2/NAP1 Fusion Protein Ag6892
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64343
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG