Learn More
Abnova™ AZGP1 (Human) Recombinant Protein
Product Code. p-7163994
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Product Code. 16122111
Brand: Abnova™ H00000563P01.10ug
Human AZGP1 full-length ORF ( AAH33830, 22 a.a. - 298 a.a.) recombinant protein with GST-tag at N-terminal.
  • Sequence: ENQGGRYSLTYIYTGLSKHVEDVPAFQALGSLNDLQFFRYNSKDRKSQPMGLWRQVEGMEDWKQDSQLQKAREDIFMETLKDIVEYYNDSNGSHVLQGRFGCEIENNRSSGAFWKYYYDGKDYIEFNKEIPAWVPFDPAAQITKQKWEAEPVYVQRAKAYLEEECPATLRKYLKYSKNILDRQDPPSVVVTSHQAPGEKKKLKCLAYDFYPGKIDVHWTRAGEVQEPELRGDVLHNGNGTYQSWVVVAVPPQDTAPYSCHVQHSSLAQPLVGPWEAS
Specifications
Accession Number AAH33830
Gene ID (Entrez) 563
Name alpha-2-glycoprotein 1, zinc-binding
Preparation Method Wheat germ expression system
Quality Control Testing 125% SDS-PAGE Stained with Coomassie Blue
Quantity 10 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias ZA2G, ZAG
Gene Symbol AZGP1
Species Wheat Germ (in vitro)
Protein Tag GST
Buffer 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer