Learn More
Azetidine-2-carboxylic acid
Product Code. 17241322 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17241322 1 g 1g
17251322 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17241322 Supplier Thermo Scientific Acros Supplier No. 466900010
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
CAS: 2517-04-6 | C4H7NO2 | 101.11 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2517-04-6
Molecular Formula C4H7NO2
Molecular Weight (g/mol) 101.11
InChI Key IADUEWIQBXOCDZ-UHFFFAOYNA-N
IUPAC Name azetidine-2-carboxylic acid
SMILES OC(=O)C1CCN1
Specifications
Quantity 1 g
Formula Weight 101.1
Chemical Name or Material Azetidine-2-carboxylic acid