missing translation for 'onlineSavingsMsg'
Learn More

AZD3839 free base, MedChemExpress

Product Code. p-200099585
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16481847

Brand: Medchem Express HY13438

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress AZD3839 free base is a potent and selective orally active, brain-permeable BACE1 inhibitor (Ki=26 nM).

AZD3839 free base shows 14 and >1000-fold selectivity against BACE2 and cathepsin D, respectively. AZD3839 free base exhibits dose- and time-dependent lowering of plasma, brain, and cerebrospinal fluid Aβ levels in mouse, guinea pig, and non-human primate. AZD3839 free base can be used for the research of Alzheimer's disease.

Specifications

Chemical Name or Material AZD3839 free base
CAS 1227163-84-9
Color White
Purity Grade Notes Research
Molecular Formula C24H16F3N5
Quantity 50 mg
Solubility Information DMSO : 125 mg/mL (289.75 mM; Need ultrasonic)
SMILES FC1=C2C([C@](C3=CC=CC(C4=CN=CN=C4)=C3)(C5=CC(C(F)F)=NC=C5)N=C2N)=CC=C1
Molecular Weight (g/mol) 431.41
Formula Weight 431.41
Percent Purity 99.89%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.