Learn More
Azaperone, MedChemExpress
Product Code. p-200096886
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Product Code. 16498766
Brand: Medchem Express HYB1470
MedChemExpress Azaperone (R-1929) acts as a dopamine antagonist but also has some antihistaminic and anticholinergic properties. Azaperone is a pyridinylpiperazine and butyrophenone neuroleptic drug with antiemetic effects, which is used mainly as a tranquilizer in veterinary medicine.
Azaperone (R-1929) acts as a dopamine antagonist but also has some antihistaminic and anticholinergic properties. Azaperone is a pyridinylpiperazine and butyrophenone neuroleptic drug with antiemetic effects, which is used mainly as a tranquilizer in veterinary medicine.
Specifications
Chemical Name or Material Azaperone
CAS 1649-18-9
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C19H22FN3O
Quantity 100 mg
Synonym R-1929
Solubility Information DMSO : 50 mg/mL (152.72 mM; Need ultrasonic) ∣H2O : 0.67 mg/mL (2.05 mM; Need ultrasonic)
SMILES O=C(C1=CC=C(F)C=C1)CCCN2CCN(C3=NC=CC=C3)CC2
Molecular Weight (g/mol) 327.4
Formula Weight 327.4
Percent Purity 99.77%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.