Learn More
Azaperone, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Azaperone |
| CAS | 1649-18-9 |
| Analyte or Component Names | Azaperone |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19 H22 F N3 O |
| Certifications/Compliance | ISO 9001 |
| Synonym | Azaperone, 1-(4-Fluorophenyl)-4-[4-(pyridin-2-yl)piperazin-1-yl]butan-1-one, 1-Butanone, 1-(4-fluorophenyl)-4-[4-(2-pyridinyl)-1-piperazinyl]-, Butyrophenone, 4'-fluoro-4-[4-(2-pyridyl)-1-piperazinyl]- (6CI,8CI), 4'-Fluoro-4-[4-(2-pyridyl)-1-piperazinyl]butyrophenone, Azaperone, Fluoperidol, NSC 170976, R-1929, Stresnil, Suicalm |
| InChI Formula | InChI=1S/C19H22FN3O/c20-17-8-6-16(7-9-17)18(24)4-3-11-22-12-14-23(15-13-22)19-5-1-2-10-21-19/h1-2,5-10H,3-4,11-15H2 |
| Solution Type | Neat |
| SMILES | Fc1ccc(cc1)C(=O)CCCN2CCN(CC2)c3ccccn3 |
| Show More |