missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AZ304, MedChemExpress
Description
AZ304 is an ATP-competitive dual BRAF kinase inhibitor, potently inhibits wild type BRAF, V600E mutant BRAF and wild type CRAF, with IC50s of 79 nM, 38 nM and 68 nM, respectively. AZ304 also has significant effect on other kinases, such as p38 (IC50, 6 nM), CSF1R (IC50, 35 nM). Anti-tumor activity.
Specifications
Specifications
| Chemical Name or Material | AZ304 |
| CAS | 942507-42-8 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C27H25N5O2 |
| Quantity | 100 mg |
| Solubility Information | DMSO : 125 mg/mL (276.84 mM; Need ultrasonic) |
| SMILES | O=C(NC1=CC=C(C)C(NC2=C3C=CC(OC)=CC3=NC=N2)=C1)C4=CC=CC(C(C)(C#N)C)=C4 |
| Molecular Weight (g/mol) | 451.52 |
| Formula Weight | 451.52 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?