missing translation for 'onlineSavingsMsg'
Learn More

AZ-23, MedChemExpress

Product Code. p-200100331
Encompass_Preferred
missing translation for 'orderingAttributeHoverText'
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
missing translation for 'unitSize'
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16492737

missing translation for 'mfr': Medchem Express HY15590

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AZ-23 is an ATP-competitive and orally bioavailable Trk kinase A/B/C inhibitor with IC50s of 2 nM (TrkA), 8 nM (TrkB), 24 nM (FGFR1), 52 nM (Flt3), 55 nM (Ret), 84 nM (MuSk), 99 nM (Lck), respectively.

AZ-23 is an ATP-competitive and orally bioavailable Trk kinase A/B/C inhibitor with IC50s of 2 nM (TrkA), 8 nM (TrkB), 24 nM (FGFR1), 52 nM (Flt3), 55 nM (Ret), 84 nM (MuSk), 99 nM (Lck), respectively.

Specifications

Chemical Name or Material AZ-23
Physical Form Solid
CAS 915720-21-7
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C17H19ClFN7O
For Use With (Application) Cancer-Kinase/protease
Synonym AZ23 AZ 23
Solubility Information DMSO : 125 mg/mL (319.02 mM; Need ultrasonic)
SMILES CC(OC1=CC(NC2=NC(N[C@H](C3=NC=C(F)C=C3)C)=NC=C2Cl)=NN1)C
Molecular Weight (g/mol) 391.83
Formula Weight 391.83
Percent Purity 98.57%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.