Learn More
AZ-23, MedChemExpress
Product Code. 16492737
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. 16492737
Brand: Medchem Express HY15590
MedChemExpress AZ-23 is an ATP-competitive and orally bioavailable Trk kinase A/B/C inhibitor with IC50s of 2 nM (TrkA), 8 nM (TrkB), 24 nM (FGFR1), 52 nM (Flt3), 55 nM (Ret), 84 nM (MuSk), 99 nM (Lck), respectively.
AZ-23 is an ATP-competitive and orally bioavailable Trk kinase A/B/C inhibitor with IC50s of 2 nM (TrkA), 8 nM (TrkB), 24 nM (FGFR1), 52 nM (Flt3), 55 nM (Ret), 84 nM (MuSk), 99 nM (Lck), respectively.
Specifications
Chemical Name or Material AZ-23
Physical Form Solid
CAS 915720-21-7
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C17H19ClFN7O
For Use With (Application) Cancer-Kinase/protease
Synonym AZ23 AZ 23
Solubility Information DMSO : 125 mg/mL (319.02 mM; Need ultrasonic)
SMILES CC(OC1=CC(NC2=NC(N[C@H](C3=NC=C(F)C=C3)C)=NC=C2Cl)=NN1)C
Molecular Weight (g/mol) 391.83
Formula Weight 391.83
Percent Purity 98.57%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.