Learn More
Axygen™ Axypet™ 8-Channel Digital Pipettor, Calibration 3x4 ALT
Shop All Axygen Scientific Products
Click to view available options
Volume (Metric):
0.5 to 10 μL
20 to 200 μL
5 to 50 μL
50 to 300 μL
Unit Size:
Each
Description
- ISO 17025 Pre-calibrated
- Thumb-friendly soft spring pipetting system reduces fatigue
- Unique side EZIject button option reduces thumb movement
- Finger-hook provides for a more relaxed grip
- Large ergonomic push button is easy on the thumb during contact
- Locking feature prevents accidental volume changes during pipetting procedures
- Push button and ejector button are ergonomically located
- Curved ejector bar allows tips to be ejected in steps, reducing the mount of force required for ejection
- Tip manifold rotates 360°
- Each channel of the pipettor has an individual, precision piston assembly to ensure accuracy and reproducibility from one pipetting series to the next
- Shafts withdraw when pressed against a row of tips
- Each shaft moves independently, minimizing jamming of tips
- Micrometer is continuously adjustable for selection of whole or fractional volumes
Specifications
Specifications
| Certifications/Compliance | ISO17025 |
| Number of Channels | 8 |
| Product Line | Axypet |
| Type | Multi-channel Pipettor |
| Volume (Metric) | 20 to 200 μL |
| Quantity | 1/Pk., 1/Cs |