Learn More
Axygen™ Axypet™ 12-Channel Digital Pipettor, Calibration 3x4 ALT
High levels of accuracy and precision with design innovations to make pipetting more comfortable during prolonged periods of time.
8850.00 NOK
Specifications
Certifications/Compliance | ISO17025 |
---|---|
Number of Channels | 12 |
Product Line | Axypet™ |
Type | Multi-channel Pipettor |
Quantity | 1/Pk., 1/Cs |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
17213614
|
Axygen™
AP-12-10-ALT |
0.5 to 10 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17273614
|
Axygen™
AP-12-50-ALT |
5 to 50 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17233614
|
Axygen™
AP-12-200-ALT |
20 to 200 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17253614
|
Axygen™
AP-12-300-ALT |
50 to 300 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- ISO 17025 Pre-calibrated
- Thumb-friendly soft spring pipetting system reduces fatigue
- Unique side EZIject button option reduces thumb movement
- Finger-hook provides for a more relaxed grip
- Large ergonomic push button is easy on the thumb during contact
- Locking feature prevents accidental volume changes during pipetting procedures
- Push button and ejector button are ergonomically located
- Curved ejector bar allows tips to be ejected in steps, reducing the mount of force required for ejection
- Tip manifold rotates 360°
- Each channel of the pipettor has an individual, precision piston assembly to ensure accuracy and reproducibility from one pipetting series to the next
- Shafts withdraw when pressed against a row of tips
- Each shaft moves independently, minimizing jamming of tips
- Micrometer is continuously adjustable for selection of whole or fractional volumes
Specifications
ISO17025 | |
Axypet™ | |
1/Pk., 1/Cs |
12 | |
Multi-channel Pipettor |