Learn More
Axygen™ AxyMats™ Sealing Mats for 500uL 96 Well Plates
Product Code. p-4879395 Shop All Axygen Scientific Products
Click to view available options
Pre-slit:
No
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 10
Pack of 50
Product Code. 11316234
Brand: Axygen™ AM500ULRD
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Minimize evaporation in wide range of temperatures in PCR and storage applications
Axygen AxyMat sealing mats can be used in a wide range of temperatures for PCR and storage applications. AxyMats minimize evaporation and are made of research-grade silicone which can be autoclaved. They can be reused following a bleach wash and ethanol rinse protocol.
TRUSTED_SUSTAINABILITY
Specifications
Color Natural
Material Silicone
Sterility Non-sterile
Autoclavable Autoclavable
No. of Wells 96
No. per Case 50/Cs.
No. per Pack 10/Pk.
Pre-slit No
Product Type Sealing Mat
Well Shape Round
Plate Compatibility 96-well Deep Well Plate, 0.5 mL