Learn More
Axygen™ Automation Micropipette Tips
Product Code. 12736679 Shop All Axygen Scientific Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Tip Style:
Automation, Genomic/Wide Orifice
Automation, Maxymum Recovery, Round
Automation, Round
Unit Size:
Pack of 4800
Pack of 960
Product Code. Sterility Tip Style unitSize
12736679 Sterile Automation, Round Pack of 4800
11384325 Sterile Automation, Round Pack of 960
12716679 Sterile Automation, Maxymum Recovery, Round Pack of 4800
11364325 Sterile Automation, Maxymum Recovery, Round Pack of 960
12033939 Non-sterile Automation, Round Pack of 4800
11374325 Non-sterile Automation, Round Pack of 960
11314335 Sterile Automation, Genomic/Wide Orifice Pack of 960
7 options
Product Code. 12736679
Brand: Axygen™ ZT200RS
Only null left
Add to basket
Add to basket
Axygen™ Automation Micropipette Tips are constructed of polypropylene, and feature a variety of available options to suit your needs.
  • For Tecan™/Freedom EVO&trade, Te-Mo™, Aquarius, Zymark™/Caliper, RapidPlate™, and SciClone™ Systems
  • For 96-well, Non-ANSI and ANSI/SLAS (384) format, number 5 polypropylene racks
  • Available with or without aerosol-resistant filters
  • Available sterile/non-sterile
  • Available in standard or Maxymum Recovery™ style
Order Info
96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 200 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Rack Color Blue
Autoclavable Autoclavable
Clean Claims RNase-free, DNase-free, Nonpyrogenic
Format Rack
Tip Style Automation, Round
Packaging 96 tips/rack, 10 racks/pack, 5 packs/case
Quantity 50 Racks/Cs.
Product Type Automation Micropipette Tip
For Use With (Equipment) Tecan/Freedom EVO, Te-Mo, Aquarius, Zymark/Caliper, RapidPlate, and SciClone Systems