Description
- Volume: 5 to 50 μL
- Clear, racked tips
- Sterile or Non-sterile
Order Info
- Microvolume tips:
- Racked: 384 tips/rack, 10 racks/pack, 5 packs/case
- Macrovolume tips:
- Bulk packaging: 250 tips/pack, 10 packs/case
Specifications
Specifications
| Volume (Metric) | 5 to 50 μL |
| Sterility | Non-sterile |
| Color | Clear |
| Autoclavable | Autoclavable |
| Clean Claims | DNase, RNase, ATP, and Endotoxin Free |
| Format | Rack |
| Tip Style | Microvolume |
| For Use With (Equipment) | Gilson Pipetman, Costar Onepette, Costar 8-Pette, Costar 12-Pette, Eppendorf, Thermo Scientific Finnpipette, Labpette Multichannel, Nichiryo, Oxford Benchmate, Rainin EDP2, Titertek |
| Material | Polypropylene |