Learn More
Axygen™ Screw Cap Tubes without Caps: Self-Standing with Conical Bottoms
Product Code. 11394225 Shop All Axygen Scientific Products
Click to view available options
Color:
Clear
Sterility:
Sterile
Capacity (Metric):
1.5 mL
2 mL
Unit Size:
Pack of 4000
Pack of 500
Product Code. 11394225
Brand: Axygen™ ST200SSS
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Self-standing tubes
Serve a broad range of laboratory needs including collection, processing, centrifugation, storage and packaging/shipping
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance RNase, DNase, pyrogen-free
Capacity (Metric) 2 mL
Color Clear
Material Polypropylene
Packaging Bag
Sterility Sterile
Closure Type Not Included
Bottom Shape Conical
Max. RCF 20,000 x g