Learn More
Axygen™ Aerosol Barrier Micropipette Tips
Shop All Axygen Scientific Products
Click to view available options
Volume (Metric):
0.5 to 50 μL
1 to 150 μL
100 to 1000 μL
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 1000
Pack of 4800
Pack of 960
Description
- Technologically advanced filter material
- Filter membrane contains no leachable additives
- Available with Maxymum Recovery™ surface for improved accuracy and maximum sample recovery
- Available in bulk, or racked (sterile)
- Available in a variety of material contact tip options
Specifications
Specifications
| Volume (Metric) | 100 to 1000 μL |
| Filtered | Filtered |
| Sterility | Non-sterile |
| Color | Clear |
| Autoclavable | Autoclavable |
| Clean Claims | DNase, RNase Free |
| Format | Bulk |
| Tip Style | Universal |
| Packaging | Bulk = 1000 tips/pack |
| Product Type | Aerosol Barrier Micropipette Tips |
| Show More |