missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Automation Micropipette Tips, PerkinElmer/MultiPROBE Janus™ Systems
GREENER_CHOICE

Product Code. 11330295 Shop All Axygen Scientific Products
Click to view available options
Volume (Metric):
50 μL
200 μL
Unit Size:
Pack of 4800
Pack of 960
This item is not returnable. View return policy

Product Code. 11330295

Brand: Axygen™ RRP50CR

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Axygen™ Automation Micropipette Tips, PerkinElmer/MultiPROBE Janus™ Systems are constructed of polypropylene, and feature a variety of available options to suit your needs.

  • Available with or without aerosol-resistant filters
  • Available sterile/non-sterile
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 50 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Clean Claims RNase-free, DNase-free, human gDNA-free, Nonpyrogenic
Format Rack
Tip Style Automation, Round
Packaging 10 racks/pk, 5 pk/cs
Product Type Automation Micropipette Tip
For Use With (Equipment) PerkinElmer MultiProbe
Product Line Tip
Material Polypropylene
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.