Specifications
| Volume (Metric) | 50 μL |
| Filtered | Non-filtered |
| Sterility | Non-sterile |
| Color | Clear |
| Format | Rack |
| Tip Style | Automation, Round |
| For Use With (Equipment) | Tecan Freedom EVO, Tecan TeMo, Tecan Aquarius MCA96 head, Zymark/Caliper RapidPlate, SciClone, Zephyr HV96, BioTek Precision, BioTek Precision XS 8, BioTek Precision XS 12, Dynamic Devices Oasis LM 8 96-Channel |
| Product Line | Tip |
| Material | Polypropylene |