Learn More
Axygen™ PlateMax™ Semi-Automated Plate Sealer Accessory, Heat Sealing Films
Shop All Axygen Scientific Products
Description
Axygen PlateMax microplate heat sealing films are designed for applications ranging from PCR and Real-time PCR to storage. They can be used in a wide range of temperatures making them suitable for nearly any application.
Specifications
Specifications
| For Use With (Application) | Plate Sealing |
| For Use With (Equipment) | PlateMax Plate Sealer |
| Material | Polyester, Aluminum |
| No. per Case | 500/Cs. |
| No. per Pack | 100/Pk. |
| Product Type | Heat Sealing Film for Compound Storage |