Learn More
Axygen™ GEN3™ Multi-Barrier Pipette Tips
Shop All Axygen Scientific Products
Click to view available options
Volume (Metric):
0.1 to 10 μL
1 to 200 μL
100 to 1000 μL
20 μL
Unit Size:
Pack of 1000
Pack of 4800
Pack of 5000
Description
The new Axygen™ brand GEN3 multi-barrier filter pipet tips offer the ultimate in protection against cross contamination using tri-filter technology. These tips will prevent liquid or aerosol from passing through and will lock on contact. GEN3 tips effectively separate liquid from the self-sealing barrier using an additional protective layer of conventional polyethylene filter material. In addition to the self-sealing properties, our GEN3 multi-barrier pipet tips feature our exclusive Maxymum Recovery™ technology which reduces the amount of reagent or sample adhering to the pipet tips. Offered in Racks, sterile, tips are nonpyrogenic and certified human gDNA and PCR inhibition free and DNase-/RNase-free.
Multi-barrier self-sealing filter prevents aerosol or liquid cross contamination Certified human gDNA and PCR inhibition, RNase-/DNase-free Nonpyrogenic Proprietary filter design prevents possible PCR inhibition Maxymum Recovery technology for ultra-low retention and high yield Sterile, no need to autoclave
Certifications
GEN3 multi-barrier tips are certified human gDNA and PCR inhibition free. Certified DNase-/RNase-free and nonpyrogenic.
Specifications
Specifications
| Volume (Metric) | 100 to 1000 μL |
| Filtered | Filtered |
| Sterility | Sterile |
| Autoclavable | Not Autoclavable |
| Clean Claims | DNase, RNase, DNA, PCR inhibitor, and Pyrogen Free |
| Format | Rack |
| Packaging | 100 Tips/Rack, 10 Racks/Pk, 5 Packs/Cs |
| Product Line | GEN3 |
| Material | Polyethylene |