Learn More
Axygen™ Conical Centrifuge Tubes
GREENER_CHOICE
Product Code. 11324175 Shop All Axygen Scientific Products
Change view
Click to view available options
Capacity (Metric):
15 mL
50 mL
Packaging:
25 Tubes and Caps/Rack, 20 Racks/Cs.
Bag
Unit Size:
Pack of 500
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Packaging unitSize
11324175 15 mL 25 Tubes and Caps/Rack, 20 Racks/Cs. Pack of 500
11354205 50 mL Bag Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11324175 Supplier Axygen™ Supplier No. SCT15ML500
Estimated Shipment: 16-12-2026
Add to basket
Add to basket
Graduated tubes with conical bottom
  • Rated to 12,000 RCF
  • Printed graduations and writing area for sample identification
  • Screw caps can be closed with one hand.
  • Sterile options are also available.
  • RNase-/DNase-free
  • Nonpyrogenic
TRUSTED_SUSTAINABILITY
Specifications
Cap Style Screw Cap
Autoclavable Not Autoclavable
Capacity (Metric) 15 mL
Color Clear
Material Polypropylene
For Use With (Application) For sample identification
Includes Screw cap
Packaging 25 Tubes and Caps/Rack, 20 Racks/Cs.
Sterility Non-sterile
Bottom Shape Conical
Max. RCF 12,000 x g