Learn More
Axygen™ Caps for PCR Tubes
Product Code. 12609535 Shop All Axygen Scientific Products
Change view
Click to view available options
Color:
Assorted
Blue
Clear
Green
MaxyClear
MaxyClear, w/writing surface
Orange
Red
Violet
Yellow
Closure Type:
Domed Cap
Flat Cap
Product Type:
8-Strip Domed PCR Tube Cap
8-Strip Flat PCR Tube Cap
8-Strip Flat PCR Tube Cap with Writing Surface
PCR Caps
Unit Size:
Pack of 100
Pack of 125
Product Code. Color Closure Type Product Type unitSize
12609535 MaxyClear, w/writing surface Flat Cap 8-Strip Flat PCR Tube Cap with Writing Surface Pack of 125
12607385 Clear Domed Cap 8-Strip Domed PCR Tube Cap Pack of 125
12547987 Blue Domed Cap 8-Strip Domed PCR Tube Cap Pack of 125
12537987 Assorted Domed Cap 8-Strip Domed PCR Tube Cap Pack of 125
12557987 Green Domed Cap 8-Strip Domed PCR Tube Cap Pack of 125
12567987 Orange Domed Cap 8-Strip Domed PCR Tube Cap Pack of 125
12587987 Violet Domed Cap 8-Strip Domed PCR Tube Cap Pack of 125
12577987 Red Domed Cap 8-Strip Domed PCR Tube Cap Pack of 125
12597987 Yellow Domed Cap 8-Strip Domed PCR Tube Cap Pack of 125
11320155 MaxyClear Flat Cap 8-Strip Flat PCR Tube Cap Pack of 125
15409073 Clear Domed Cap PCR Caps Pack of 100
11 options
Product Code. 12609535
Brand: Axygen™ PCR02FCPC
8-Strip domed or flat strip caps for Axygen PCR tubes
Axygen polypropylene, thin wall PCR tubes with and without caps are designed to fit standard 96-well blocks and are available in an assortment of colors. The consistent and ultra-thin wall allows precise thermal transfer. An even insertion cap ensures a tight fit to reduce evaporation.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance RNase, DNase, pyrogen-free
Closure Color Clear
Color MaxyClear, w/writing surface
Volume (Metric) 0.2 mL
Quantity 1250/Cs.
Closure Material Polypropylene
For Use With (Equipment) 8-strips PCR tubes and 96-well PCR microplates
Sterility Non-sterile
Material Polypropylene
Closure Type Flat Cap
Format Strip
Product Type 8-Strip Flat PCR Tube Cap with Writing Surface