missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Biomek™ FX/Wellpro™ Robotic Tips
GREENER_CHOICE

Product Code. 11306304 Shop All Axygen Scientific Products
Click to view available options
Sterility:
Non-sterile
Sterile
Tip Style:
Automation, Extended Length
Extended Length, Automation
Low Retention, Extended Length, Automation
Unit Size:
Pack of 19200
Pack of 3840
This item is not returnable. View return policy

Product Code. 11306304

Brand: Axygen™ FX384XLRS

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For Beckman FX/Wellpro system

  • Tip format: 384 tip
  • Tip volume range: 25 μL to 50 μL
  • Tip material: Clear polypropylene
  • RNase-/DNase-/human gDNA-free
  • Nonpyrogenic
  • Available options:
    - With or without aerosol-resistant filters
    - Nonsterile or sterile 
    - Standard or Axygen™ Maxymum Recovery™ surface
  • Compatible head: AP-384, 384-channel low volume (FXF-384-XL-R-S only)
  • Rack format: ANSI/SLAS
  • Rack material: Polypropylene
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 50 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Autoclavable Not Autoclavable
Clean Claims DNase, RNase and Pyrogen free
For Use With (Application) Non-pyrogenic, used for stringent specifications under strict process controls
Format Rack, 384-well
Tip Style Extended Length, Automation
Packaging 10 Racks/Pk, 5 Pks/Cs.
Quantity 50 Racks/Cs.
For Use With (Equipment) Beckman Coulter Biomek FX, Beckman Coulter Biomek NX, Beckman Coulter Multimek AP384, Dynamic Devices Oasis, Dynamic Devices Oasis LM 384-Channel Low Volume, Hamilton
Product Line Beckman Biomek FX/Wellpro
Material Polypropylene
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.