Learn More
Axygen™ Biomek™ FX/NX Robotic Tips
GREENER_CHOICE
Product Code. 12585727 Shop All Axygen Scientific Products
Change view
Click to view available options
Volume (Metric):
10 μL
20 μL
30 μL
45 μL
50 μL
250 μL
Sterility:
Non-sterile
Sterile
Tip Style:
Automation, Fine Point
Automation, Round
Automation, Wide Orifice
Unit Size:
Pack of 19200
Pack of 3840
Pack of 4800
Pack of 960
Product Code. Volume (Metric) Sterility Tip Style unitSize
12585727 30 μL Sterile Automation, Round Pack of 3840
11316294 10 μL Non-sterile Automation, Round Pack of 960
12732846 10 μL Non-sterile Automation, Round Pack of 4800
12742846 10 μL Sterile Automation, Round Pack of 4800
11326294 10 μL Sterile Automation, Round Pack of 960
11980948 20 μL Non-sterile Automation, Round Pack of 960
12792846 20 μL Non-sterile Automation, Round Pack of 4800
12702856 20 μL Sterile Automation, Round Pack of 4800
12519177 20 μL Sterile Automation, Round Pack of 960
12772846 20 μL Non-sterile Automation, Round Pack of 4800
12599167 20 μL Non-sterile Automation, Round Pack of 960
12509177 20 μL Sterile Automation, Round Pack of 960
12782846 20 μL Sterile Automation, Round Pack of 4800
11346294 30 μL Sterile Automation, Fine Point Pack of 3840
15591383 30 μL Non-sterile Automation, Fine Point Pack of 19200
12722866 30 μL Sterile Automation, Round Pack of 19200
12712866 30 μL Non-sterile Automation, Round Pack of 19200
12575727 30 μL Non-sterile Automation, Round Pack of 3840
15501393 30 μL Sterile Automation, Fine Point Pack of 19200
12792856 30 μL Non-sterile Automation, Round Pack of 19200
11356294 30 μL Non-sterile Automation, Round Pack of 3840
12772856 45 μL Non-sterile Automation, Fine Point Pack of 19200
12772866 50 μL Non-sterile Automation, Round Pack of 4800
12579177 50 μL Non-sterile Automation, Round Pack of 960
12782866 50 μL Sterile Automation, Round Pack of 4800
12589177 50 μL Sterile Automation, Round Pack of 960
12792866 50 μL Non-sterile Automation, Round Pack of 4800
12599177 50 μL Non-sterile Automation, Round Pack of 960
12509187 50 μL Sterile Automation, Round Pack of 960
12702876 50 μL Sterile Automation, Round Pack of 4800
12712856 250 μL Non-sterile Automation, Round Pack of 4800
12529177 250 μL Non-sterile Automation, Round Pack of 960
12722856 250 μL Sterile Automation, Round Pack of 4800
12539177 250 μL Sterile Automation, Round Pack of 960
11990948 250 μL Non-sterile Automation, Round Pack of 960
12732856 250 μL Non-sterile Automation, Round Pack of 4800
12742856 250 μL Sterile Automation, Round Pack of 4800
12549177 250 μL Sterile Automation, Round Pack of 960
12752856 250 μL Non-sterile Automation, Wide Orifice Pack of 4800
12559177 250 μL Non-sterile Automation, Wide Orifice Pack of 960
12762856 250 μL Sterile Automation, Wide Orifice Pack of 4800
12569177 250 μL Sterile Automation, Wide Orifice Pack of 960
42 options
Product Code. 12585727
Brand: Axygen™ FX384RS
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Axygen robotic tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality.
The Axygen brand range of products includes a comprehensive line of tips for Beckman Coulter automated liquid handling systems. These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality. The pipette tips are also available with MAXYMum Recovery, a unique low-retention technology that improves tip performance and maximizes the use of your precious samples.
  • Available in racked, racked sterile and with aerosol barrier (filtered)
  • Compatible with Beckman Biomek FX and Beckman Multimek instruments with an AP96 head
  • Also compatible with Span-8 pod when Liquid Level Sensing function is turned off
  • Unsurpassed straightness to ensure tip enters the well smoothly, every time
  • 384/1536-well compatible to meet your most demanding applications
  • Wide bore tips are ideal for pipetting viscous materials, genomic DNA or delicate cell cultures
  • Fine Point improves accuracy and precision at lower sample volumes
  • Maxymum recovery™ tips reduce sample loss, improves reproducibility and accuracy, minimizes usage of precious samples and reagents
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 30 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Autoclavable Not Autoclavable
Certifications/Compliance ANSI
Clean Claims DNase, RNase and Pyrogen free
For Use With (Application) Non-pyrogenic, used for stringent specifications under strict process controls
Format Rack, 384-well
Tip Style Automation, Round
Packaging 10 Racks/Pk, 5 Pks/Cs.
For Use With (Equipment) Beckman Coulter Biomek FX, Beckman Coulter Biomek NX, Beckman Coulter Biomek 3000, Beckman Coulter Multimek AP96, Beckman Coulter Multimek SPAN-8, Beckman Coulter Multimek MP20, Beckman Coulter Multimek MP200
Product Line Beckman Biomek