Learn More
Axygen™ AxyMats™ Sealing Mats for 384 Well Deep Well Plates
Minimize evaporation in wide range of temperatures in PCR and storage applications
Brand: Axygen™ AM-384-PCR-RD
Additional Details : Weight : 0.82000kg
Description
Axygen AxyMat sealing mats can be used in a wide range of temperatures for PCR and storage applications. AxyMats minimize evaporation and are made of research-grade silicone which can be autoclaved. They can be reused following a bleach wash and ethanol rinse protocol.
Specifications
Clear | |
Non-sterile | |
384 | |
10/Pk. | |
Round |
Silicone | |
Autoclavable | |
50/Cs. | |
Sealing Mat | |
384-well Deep Well Plate with Round Wells |