Learn More
Axygen™ AxyMats™ Sealing Mats for 384 Well Deep Well Plates
Minimize evaporation in wide range of temperatures in PCR and storage applications
Brand:  Axygen™ AM-384-DW-SQ
Additional Details : Weight : 1.22000kg
 View more versions of this product
Product Code. 11376224
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Axygen AxyMat sealing mats can be used in a wide range of temperatures for PCR and storage applications. AxyMats minimize evaporation and are made of research-grade silicone which can be autoclaved. They can be reused following a bleach wash and ethanol rinse protocol.
Specifications
Specifications
Natural
Non-sterile
384
10/Pk.
Square
Silicone
Autoclavable
50/Cs.
Sealing Mat
384-well Deep Well Plate with Square Wells