Learn More
Axygen™ AxyMats™ Pierceable Sealing Mats for 24 Well Deep Well Plates
Minimize evaporation in wide range of temperatures in PCR and storage applications.
Brand: Axygen™ AM-24-SQ
Additional Details : Weight : 1.41300kg
Description
- Axygen AxyMat sealing mats can be used in a wide range of temperatures for PCR and storage applications.
- AxyMats minimize evaporation and are made of research-grade silicone which can be autoclaved.
- They can be reused following a bleach wash and ethanol rinse protocol.
- Mfr. No. AM-24-SQ
Specifications
Silicone | |
Autoclavable | |
50/Cs. | |
Sealing Mat | |
24-well Deep Well Plate with Square Wells |
Non-sterile | |
24 | |
10/Pk. | |
Square |