Learn More
Axygen™ AxyMats™ Pierceable Sealing Mats for 24 Well Deep Well Plates
Minimize evaporation in wide range of temperatures in PCR and storage applications.
Brand: Axygen™ AM-24-SQ-S
Additional Details : Weight : 1.13000kg
Description
- Axygen AxyMat sealing mats can be used in a wide range of temperatures for PCR and storage applications
- AxyMats minimize evaporation and are made of research-grade silicone which can be autoclaved
- They can be reused following a bleach wash and ethanol rinse protocol.
- Mfr. No. AM-24-SQ-S
Specifications
Silicone | |
Autoclavable | |
5 | |
Sealing Mat |
Sterile | |
24 | |
10 | |
24-well Deep Well Plate with Square Wells |