Learn More
Axygen™ 24- and 32-Well PCR Microplates, Clear, Nonsterile
GREENER_CHOICE
Product Code. 11310155 Shop All Axygen Scientific Products
Change view
Click to view available options
No. of Wells:
24
32
Unit Size:
Pack of 10
Pack of 50
Product Code. No. of Wells unitSize
11310155 24 Pack of 10
12789408 24 Pack of 50
11330155 32 Pack of 10
12799408 32 Pack of 50
4 options
Product Code. 11310155
Brand: Axygen™ PCR24C
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Designed to fit a wide variety of thermal cyclers
  • Volume to brim of well: 350 μL, working volume: 300 μL, volume with AxyMat: 240 μL
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance RNase, DNase-free
Format Standard
Lid Without Lid
Array 4 x 6
Color Clear
Product Type PCR Microplate, without Lid
Material Polypropylene
No. of Wells 24
Sterility Non-sterile
Well Shape Round
For Use With (Application) PCR
No. per Case 50/Cs.
No. per Pack 10/Pk.