Learn More
Axygen™ 200 μL Universal Pipetter Tips: 200 μL, Wide Bore
Product Code. 11396054 Shop All Axygen Scientific Products
Click to view available options
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 1000
Pack of 4800
Product Code. 11396054
Brand: Axygen™ T205WBCRS
Only null left
Add to basket
Add to basket
Designed to fit a wide variety of single and multi-channel pipettes
Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.
(This is the suggested replacement for T-205-WB-C-L)
  • These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.
  • Nonpyrogenic
  • Wide-Bore
Order Info
Available in bulk, racked or stack rack formats.
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 1 to 200 μL
Sterility Sterile
Color Clear
Autoclavable Autoclavable
Clean Claims DNase, RNase, and Pyrogen free
Format Rack
Tip Style Genomic/Wide Orifice, Universal
For Use With (Equipment) Universal Fit