Learn More
Axygen™ Maxymum Recovery™ 20 μL Ultra Micro Tips
Maxymum Recovery™ allows recovery of costly samples with improved precision and accuracy
Brand: Axygen™ T-400-L-R
Additional Details : Weight : 7.71000kg
Description
Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors, compatible with most popular brands of pipettors. These polypropylene tips are RNase-/DNase-free and nonpyrogenic. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.
- These polypropylene tips are nonpyrogenic andRNase-/DNase-free.
- Nonpyrogenic
- Maxymum Recovery technology for ultra-low retention and high yield
Specifications
0.5 to 20 μL | |
Clear | |
Allows recovery of costly samples with improved precision and accuracy | |
MicroPoint, Low Retention | |
Eppendorf Pipetter, Gilson™ Pipetman™ P2, Gilson™ Pipetman™ P10 | |
Polypropylene |
Non-sterile | |
DNase, RNase Free | |
Rack | |
96 Tips/Rack, 10 Racks/Pk, 5 Packs/Cs | |
Maximum Recovery |