Learn More
Axygen™ 20 μL Ultra Micro Tips
Product Code. 12726619 Shop All Axygen Scientific Products
Click to view available options
Sterility:
Non-sterile
Sterile
Format:
Bulk
Rack
Unit Size:
Pack of 1000
Pack of 20000
Pack of 4800
Pack of 960
Product Code. 12726619
Brand: Axygen™ T400RS
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Maxymum Recovery™ allows recovery of costly samples with improved precision and accuracy
Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors, compatible with most popular brands of pipettors. These polypropylene tips are nonpyrogenic and RNase-/DNase-free. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.
  • RNase-/DNase-free
  • Nonpyrogenic
  • Maxymum Recovery technology for ultra-low retention
Order Info
  • Bulk: 1000 tips/pack, 20 packs/case
  • Racked: 96 tips/rack, 10 racks/pack, 5 packs/case
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 20 μL
Sterility Sterile
Color Clear
Clean Claims DNase, RNase Free
For Use With (Application) Allows recovery of costly samples with improved precision and accuracy
Format Rack
Tip Style Micro Point
For Use With (Equipment) Eppendorf, Gilson Pipetman P2, Gilson Pipetman P10