Learn More
Axygen™ 20 μL Ultra Micro Tips
Product Code. 12629535 Shop All Axygen Scientific Products
Click to view available options
Sterility:
Non-sterile
Sterile
Format:
Bulk
Rack
Unit Size:
Pack of 1000
Pack of 20000
Pack of 4800
Pack of 960
Product Code. 12629535
Brand: Axygen™ T400R
Only null left
Add to basket
Add to basket
Maxymum Recovery™ allows recovery of costly samples with improved precision and accuracy
Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors, compatible with most popular brands of pipettors. These polypropylene tips are nonpyrogenic and RNase-/DNase-free. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.
  • RNase-/DNase-free
  • Nonpyrogenic
  • Maxymum Recovery technology for ultra-low retention
Order Info
  • Bulk: 1000 tips/pack, 20 packs/case
  • Racked: 96 tips/rack, 10 racks/pack, 5 packs/case
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 0.5 to 20 μL
Sterility Non-sterile
Color Clear
Clean Claims DNase, RNase Free
For Use With (Application) Allows recovery of costly samples with improved precision and accuracy
Format Rack
Tip Style Micro Point
Packaging 96 Tips/Rack, 10 Racks/Pk, 5 Packs/Cs
For Use With (Equipment) Eppendorf, Gilson Pipetman P2, Gilson Pipetman P10