Learn More
Carl Zeiss™ Axio Vert.A1 Inverted Microscope
Product Code. 12818065
Change view
Click to view available options
Lighting Type:
Halogen
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Lighting Type unitSize
12818065 Halogen Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12818065 Supplier Carl Zeiss™ Supplier No. 4912370002000
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Compact inverted microscope with all standard contrast technique options. Carl Zeiss™ Axio Vert.A1 Inverted Microscope features ergonomic design of all operating parts.
  • Versatile: all standard contrasting techniques including DIC
  • Transmitted-light brightfield HAL
  • Phase contrast (Ph1)
  • Suitable for a variety of biomedical and materials applications
Specifications
Includes Dust protection set
Condenser Type LD condenser 0.4 for slider
For Use With (Application) A variety of applications benefiting from contrast techniques.
Eyepiece PL 10x/23 Br. Foc.
Lighting Type Halogen
Product Type Inverted Microscope