Learn More
AVE 0991 sodium salt, MedChemExpress
Product Code. 16492897
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16492897
Brand: Medchem Express HY15778A
MedChemExpress AVE 0991 sodium salt is a nonpeptide and orally active Ang-(1-7) receptor Mas agonist. AVE 0991 competes for high-affinity binding of [125I]-Ang-(1-7) to bovine aortic endothelial cell membranes with IC50 of 21 nM.
AVE 0991 sodium salt is a nonpeptide and orally active Ang-(1-7) receptor Mas agonist. AVE 0991 competes for high-affinity binding of [125I]-Ang-(1-7) to bovine aortic endothelial cell membranes with IC50 of 21 nM.
Specifications
Chemical Name or Material AVE 0991 sodium salt
CAS 306288-04-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C29H31N4NaO5S2
Quantity 50 mg
Solubility Information DMSO : ≥ 55 mg/mL (91.26 mM) ∣H2O : 50 mg/mL (82.96 mM; Need ultrasonic)
SMILES O=S(C1=C(C2=CC=C(CN3C(C=O)=C(OC)N=C3C4=CC=CC=C4)C=C2)C=C(CC(C)C)S1)([N-]C(NCC)=O)=O.[Na+]
Molecular Weight (g/mol) 602.7
Formula Weight 602.7
Percent Purity 98.28%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.