missing translation for 'onlineSavingsMsg'
Learn More

AVE 0991 sodium salt, MedChemExpress

Product Code. 16476828
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16476828

Brand: Medchem Express HY15778A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AVE 0991 sodium salt is a nonpeptide and orally active Ang-(1-7) receptor Mas agonist. AVE 0991 competes for high-affinity binding of [125I]-Ang-(1-7) to bovine aortic endothelial cell membranes with IC50 of 21 nM.

AVE 0991 sodium salt is a nonpeptide and orally active Ang-(1-7) receptor Mas agonist. AVE 0991 competes for high-affinity binding of [125I]-Ang-(1-7) to bovine aortic endothelial cell membranes with IC50 of 21 nM.

Specifications

Chemical Name or Material AVE 0991 sodium salt
CAS 306288-04-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C29H31N4NaO5S2
Quantity 10 mg
Solubility Information DMSO : ≥ 55 mg/mL (91.26 mM) ∣H2O : 50 mg/mL (82.96 mM; Need ultrasonic)
SMILES O=S(C1=C(C2=CC=C(CN3C(C=O)=C(OC)N=C3C4=CC=CC=C4)C=C2)C=C(CC(C)C)S1)([N-]C(NCC)=O)=O.[Na+]
Molecular Weight (g/mol) 602.7
Formula Weight 602.7
Percent Purity 98.28%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.