missing translation for 'onlineSavingsMsg'
Learn More

AV-412 free base, MedChemExpress

Product Code. 16410227
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16410227 50 mg 50mg
16483848 10 mg 10mg
16436577 5 mg 5mg
16434886 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16410227 Supplier Medchem Express Supplier No. HY10346A
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress AV-412 free base (MP-412 free base) is an EGFR inhibitor with IC50s of 0.75, 0.5, 0.79, 2.3, 19 nM for EGFR, EGFRL858R, EGFRT790M, EGFRL858R/T790M and ErbB2, respectively.

AV-412 free base (MP-412 free base) is an EGFR inhibitor with IC50s of 0.75, 0.5, 0.79, 2.3, 19 nM for EGFR, EGFRL858R, EGFRT790M, EGFRL858R/T790M and ErbB2, respectively.

Specifications

Chemical Name or Material AV-412 free base
CAS 451492-95-8
Color Yellow
Purity Grade Notes Research
Molecular Formula C27H28ClFN6O
Quantity 50 mg
Synonym MP-412 free base
Solubility Information DMSO : 50 mg/mL (98.62 mM; Need ultrasonic)
SMILES O=C(C=C)NC1=CC2=C(NC3=CC=C(C(Cl)=C3)F)N=CN=C2C=C1C#CC(C)(C)N(CC4)CCN4C
Molecular Weight (g/mol) 507.0
Formula Weight 507.0
Percent Purity 98.33%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.