Learn More
Trajan™ Autosampler Syringes With Removable Needle for Thermo Scientific GCs
Product Code. 13169195
Change view
Click to view available options
Needle Gauge:
23 G
26 G
Diameter (English) Outer Needle:
0.024 in.
0.18 in.
0.24 in.
Length (English) Needle:
2.24 in.
3.34 in.
Model:
0.5BR-RTC/RSH-5.7/0.63C
10R-RTC/RSH-5.7/0.47C
10R-RTC/RSH-5.7/0.63C
10R-RTC/RSH-GT-5.7/0.47C
10R-RTC/RSH-GT-5.7/0.63C
10R-RTC/RSH-GT-8.5/0.47C
10R-RTC/RSH-GT-8.5/0.63C
1BR-RTC/RSH-5.7/0.63C
5R-RTC/RSH-5.7/0.47C
5R-RTC/RSH-5.7/0.63C
5R-RTC/RSH-8.5/0.47C
Unit Size:
Each
Product Code. Needle Gauge Diameter (English) Outer Needle Length (English) Needle Model unitSize
13169195 23 G 0.024 in. 2.24 in. 1BR-RTC/RSH-5.7/0.63C Each
13159015 26 G 0.18 in. 3.34 in. 5R-RTC/RSH-8.5/0.47C Each
13159195 23 G 0.024 in. 2.24 in. 0.5BR-RTC/RSH-5.7/0.63C Each
13126115 23 G 0.024 in. 2.24 in. 5R-RTC/RSH-5.7/0.63C Each
13149205 26 G 0.18 in. 3.34 in. 10R-RTC/RSH-GT-8.5/0.47C Each
13149015 26 G 0.18 in. 2.24 in. 5R-RTC/RSH-5.7/0.47C Each
13129205 23 G 0.24 in. 3.34 in. 10R-RTC/RSH-GT-8.5/0.63C Each
13119205 23 G 0.24 in. 2.24 in. 10R-RTC/RSH-GT-5.7/0.63C Each
13129025 26 G 0.18 in. 2.24 in. 10R-RTC/RSH-5.7/0.47C Each
13109025 23 G 0.24 in. 2.24 in. 10R-RTC/RSH-5.7/0.63C Each
13139205 26 G 0.18 in. 2.24 in. 10R-RTC/RSH-GT-5.7/0.47C Each
11 options
Product Code. 13169195
Brand: Trajan™ 000680
For reduced non specific sample interaction and increased analyte recovery
  • Borosilicate glass barrel for strength and inertness.
  • Stainless steel needles for inertness, strength and corrosion resistance.
  • Backing behind the scale aids quick and accurate reading of the syringe scale.
  • Metal back flange provides stability and resists fracture.
  • Metal needle cap on syringe barrel protects and improves syringe lifetime.
  • Syringes are individually tested for quality ensuring reliability and conformance.
  • Unique packaging design eliminates possible contamination in the syringe.
Specifications
Needle Gauge 23 G
Diameter (English) Outer Needle 0.024 in.
Diameter (Metric) Outer Needle 0.63 mm
For Use With (Application) Gas chromatography
Length (English) Needle 2.24 in.
Product Type Autosampler Syringe
Model 1BR-RTC/RSH-5.7/0.63C
Quantity Ea.
Volume (Metric) 1 μL
Needle Point Style Cone
For Use With (Equipment) Thermo GC, CTC PAL RTC (Robotic Tool Change) Autosampler
Connection Type Removable