Learn More
Trajan™ Autosampler Syringes with Removable Needle for CTC Analytics CombiPAL and GC PAL
Product Code. 13186105
Change view
Click to view available options
Needle Gauge:
23 G
26 G
Diameter (English) Outer Needle:
0.01 in.
0.018 in.
0.02 in.
Length (English) Needle:
1.9 in.
2.2 in.
3.3 in.
Model:
10F-RTC/RSH-5.7/0.47C
10F-RTC/RSH-5.7/0.63C
10F-RTC/RSH-8.5/0.47C
10F-RTC/RSH-8.5/0.63C
10F-RTC/RSH-GT-5.7/0.47C
10F-RTC/RSH-GT-5.7/0.63C
10F-RTC/RSH-GT-8.5/0.47C
10F-RTC/RSH-GT-8.5/0.63C
25F-RTC/RSH-GT-5.7/0.47C
25F-RTC/RSH-GT-5.7/0.63C
25F-RTC/RSH-GT-8.5/0.63C
25FRTC/RSH-GT-8.5/0.47C
25R-C/T-GT-0.47
50F-RTC/RSH-GT-5.7/0.47C
50F-RTC/RSH-GT-5.7/0.63C
50F-RTC/RSH-GT-8.5/0.47C
50F-RTC/RSH-GT-8.5/0.63C
5F-RTC/RSH-5.7/0.47C
5F-RTC/RSH-5.7/0.63C
5F-RTC/RSH-8.5/0.63C
Unit Size:
Each
Product Code. Needle Gauge Diameter (English) Outer Needle Length (English) Needle Model unitSize
13186105 23 G 0.02 in. 2.2 in. 5F-RTC/RSH-5.7/0.63C Each
13189015 26 G 0.01 in. 2.2 in. 10F-RTC/RSH-5.7/0.47C Each
13199195 26 G 0.01 in. 2.2 in. 10F-RTC/RSH-GT-5.7/0.47C Each
13179015 23 G 0.02 in. 3.3 in. 10F-RTC/RSH-8.5/0.63C Each
13109205 26 G 0.01 in. 3.3 in. 10F-RTC/RSH-GT-8.5/0.47C Each
13109215 23 G 0.02 in. 3.3 in. 50F-RTC/RSH-GT-8.5/0.63C Each
13106115 26 G 0.01 in. 2.2 in. 5F-RTC/RSH-5.7/0.47C Each
13169015 23 G 0.02 in. 2.2 in. 10F-RTC/RSH-5.7/0.63C Each
13196105 23 G 0.02 in. 3.3 in. 5F-RTC/RSH-8.5/0.63C Each
13129215 26 G 0.01 in. 3.3 in. 50F-RTC/RSH-GT-8.5/0.47C Each
13199205 23 G 0.02 in. 2.2 in. 50F-RTC/RSH-GT-5.7/0.63C Each
13179195 23 G 0.02 in. 2.2 in. 10F-RTC/RSH-GT-5.7/0.63C Each
13189205 26 G 0.01 in. 3.3 in. 25FRTC/RSH-GT-8.5/0.47C Each
13179205 26 G 0.01 in. 2.2 in. 25F-RTC/RSH-GT-5.7/0.47C Each
13119215 26 G 0.01 in. 2.2 in. 50F-RTC/RSH-GT-5.7/0.47C Each
13189195 23 G 0.02 in. 3.3 in. 10F-RTC/RSH-GT-8.5/0.63C Each
13159205 23 G 0.02 in. 2.2 in. 25F-RTC/RSH-GT-5.7/0.63C Each
13169205 23 G 0.02 in. 3.3 in. 25F-RTC/RSH-GT-8.5/0.63C Each
13199015 26 G 0.01 in. 3.3 in. 10F-RTC/RSH-8.5/0.47C Each
12977506 26 G 0.018 in. 1.9 in. 25R-C/T-GT-0.47 Each
20 options
Product Code. 13186105
Brand: Trajan™ 001861
For reduced non specific sample interaction and increased analyte recovery
  • Borosilicate glass barrel for strength and inertness.
  • Stainless steel needles for inertness, strength and corrosion resistance.
  • Backing behind the scale aids quick and accurate reading of the syringe scale.
  • Metal back flange provides stability and resists fracture.
  • Metal needle cap on syringe barrel protects and improves syringe lifetime.
  • Syringes are individually tested for quality ensuring reliability and conformance.
  • Unique packaging design eliminates possible contamination in the syringe.
Specifications
Needle Gauge 23 G
Diameter (English) Outer Needle 0.02 in.
Diameter (Metric) Outer Needle 0.63 mm
For Use With (Application) Gas chromatography
Length (English) Needle 2.2 in.
Product Type Autosampler Syringe
Model 5F-RTC/RSH-5.7/0.63C
Quantity Ea.
Volume (Metric) 5 μL
Needle Point Style Cone
For Use With (Equipment) Thermo RSH Autosampler
Connection Type Fixed